4-Chloro-3-nitrophenyl isocyanate

Supplier: APOLLO SCIENTIFIC
Danger

OR0268-1G OR0268-5G
77401-980EA 33.62 USD
77401-980 77401-982
4-Chloro-3-nitrophenyl isocyanate
4-Chloro-3-nitrophenyl isocyanate

When melted, may contain small amount of sediment.

Formula: C₇H₃ClN₂O₃
Melting Pt: 63…69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037062
CAS Number: 40397-96-4
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR