4-(Chloromethyl)-3,5-dimethylisoxazole

Supplier: APOLLO SCIENTIFIC
Warning

OR0256-25G OR0256-5G OR0256-100G
77401-974EA 58.08 USD
77401-974 77401-976 77401-972
4-(Chloromethyl)-3,5-dimethylisoxazole
4-(Chloromethyl)-3,5-dimethylisoxazole
Formula: C₆H₈ClNO
MW: 145.59 g/mol
Boiling Pt: 88…90 °C/7 mmHg
Density: 1.167 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003154
CAS Number: 19788-37-5
EINECS: 243-312-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR