5-Chloro-2-nitrobenzamide

Supplier: APOLLO SCIENTIFIC
Warning

OR0028-100G OR0028-25G OR0028-5G
77401-642EA 268.96 USD
77401-642 77401-644 77401-646
5-Chloro-2-nitrobenzamide
5-Chloro-2-nitrobenzamide
Formula: C₇H₅ClN₂O₃
MW: 200.58 g/mol
Melting Pt: 157…160 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00017509
CAS Number: 40763-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR