3-Amino-1-ethyl-1H-pyrazole

Supplier: APOLLO SCIENTIFIC
Warning

OR111552-5G OR111552-10G OR111552-1G
77403-814EA 323.98 USD
77403-814 77403-810 77403-812
3-Amino-1-ethyl-1H-pyrazole
3-Amino-1-ethyl-1H-pyrazole
Formula: C₅H₉N₃
Storage Temperature: Ambient
MDL Number: MFCD03422571
CAS Number: 55361-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR