Ethyl 3-amino-7-methoxy-1-benzofuran-2-carboxylate

Supplier: APOLLO SCIENTIFIC

OR111382-1G OR111382-5G
77403-742EA 212.94 USD
77403-742 77403-744
Ethyl 3-amino-7-methoxy-1-benzofuran-2-carboxylate
Ethyl 3-amino-7-methoxy-1-benzofuran-2-carboxylate
Formula: C₁₂H₁₃NO₄
MW: 235.236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526241
CAS Number: 1351479-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR