(4S,5R)-(-)-4-Methyl-5-phenyl-2-oxazolidinone

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (4S,5R)-4-Methyl-5-phenyloxazolidin-2-one, (4S,5R)-(-)-4-Methyl-5-phenyloxazolidin-2-one

OR10970-1G OR10970-250MG OR10970-5G
77403-006EA 55.02 USD
77403-006 77403-008 77403-010
(4S,5R)-(-)-4-Methyl-5-phenyl-2-oxazolidinone
(4S,5R)-(-)-4-Methyl-5-phenyl-2-oxazolidinone
Formula: C₁₀H₁₁NO₂
MW: 177.2000 g/mol
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00066226
CAS Number: 16251-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR