Dimethyl (2-oxopropyl)phosphonate 97+%

Supplier: APOLLO SCIENTIFIC
Warning

OR10776-100G OR10776-25G OR10776-5G
77402-918EA 206.82 USD
77402-918 77402-920 77402-922
Dimethyl (2-oxopropyl)phosphonate 97+%
Dimethyl (2-oxopropyl)phosphonate
Formula: C₅H₁₁O₄P
Boiling Pt: 76…79 °C/3 mmHg
Density: 1.202 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008769
CAS Number: 4202-14-6
EINECS: 224-110-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR