(3-Ethoxy-3-oxopropyl)(triphenyl)phosphonium bromide 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (3-Ethoxy-3-oxopropyl)triphenylphosphonium bromide

OR10644-1G OR10644-25G OR10644-5G
77402-852EA 23.44 USD
77402-852 77402-854 77402-856
(3-Ethoxy-3-oxopropyl)(triphenyl)phosphonium bromide 97%
(3-Ethoxy-3-oxopropyl)(triphenyl)phosphonium bromide
Formula: C₂₃H₂₄O₂P·Br
MW: 443.313 g/mol
Melting Pt: 145 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD03701355
CAS Number: 42843-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR