4,5-Dichlorophthalonitrile 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR1063-5G OR1063-1G OR1063-250MG
77402-834EA 481.9 USD
77402-834 77402-830 77402-832
4,5-Dichlorophthalonitrile 95%
4,5-Dichlorophthalonitrile
Formula: C₈H₂Cl₂N₂
Melting Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00191408
CAS Number: 139152-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR