7-Hydroxyindole 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR10209-1G OR10209-5G
77402-544EA 110.04 USD
77402-544 77402-546
7-Hydroxyindole 97%
7-Hydroxyindole
Formula: C₈H₇NO
MW: 133.15 g/mol
Melting Pt: 84…88 °C
Storage Temperature: Freezer
MDL Number: MFCD00152102
CAS Number: 2380-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR