Indole-5-carbonitrile

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1H-Indole-5-carbonitrile, 5-Cyanoindole, 5-Cyano-1H-indole

OR10181-5G OR10181-25G
77402-534EA 21.32 USD
77402-534 77402-532
Indole-5-carbonitrile
Indole-5-carbonitrile
Formula: C₉H₆N₂
MW: 142.16 g/mol
Melting Pt: 106…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00005669
CAS Number: 15861-24-2
EINECS: 239-986-4
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR