3-Amino-4-pyrazolecarbonitrile

Supplier: APOLLO SCIENTIFIC
Warning

OR0352T-100G OR0352T-250G OR0352T-25G
77402-046EA 41.78 USD
77402-046 77402-048 77402-050
3-Amino-4-pyrazolecarbonitrile
3-Amino-4-pyrazolecarbonitrile

A synthetic intermediate.

Formula: C₄H₄N₄
Melting Pt: 172…176 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005237
CAS Number: 16617-46-2
EINECS: 240-665-6
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR