3-(tert-Butyl)phenylhydrazine hydrochloride, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (3-(tert-Butyl)phenyl)hydrazine hydrochloride, 1-(tert-Butyl)-3-hydrazinobenzene

OR016187-1G OR016187-250MG OR016187-5G
77401-782EA 206.82 USD
77401-782 77401-784 77401-786
3-(tert-Butyl)phenylhydrazine hydrochloride, Technical Grade
3-(tert-Butyl)phenylhydrazine hydrochloride
Formula: C₁₀H₁₆N₂·ClH
Melting Pt: 146…148 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00833404
CAS Number: 306937-27-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR