Phenyl(phenylamino)acetic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR01496-1G OR01496-250MG
77401-712EA 290.36 USD
77401-712 77401-714
Phenyl(phenylamino)acetic acid 95%
Phenyl(phenylamino)acetic acid
Formula: C₁₄H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD01724775
CAS Number: 3684-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR