7-Aminopyrazolo[1,5-a]pyrimidine-6-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 7-Amino-6-carboxypyrazolo[1,5-a]pyrimidine

OR103647-1G OR103647-250MG
77402-634EA 303.6 USD
77402-634 77402-636
7-Aminopyrazolo[1,5-a]pyrimidine-6-carboxylic acid
7-Aminopyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Formula: C₇H₆N₄O₂
MW: 178.148 g/mol
Melting Pt: 242 °C
Storage Temperature: Ambient
MDL Number: MFCD00973861
CAS Number: 68262-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR