N,N-Dimethyl2-piperazin-1-ylacetamide 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR0239-10G OR0239-1G OR0239-5G
77401-956EA 294.44 USD
77401-956 77401-958 77401-960
N,N-Dimethyl2-piperazin-1-ylacetamide 97%
N,N-Dimethyl2-piperazin-1-ylacetamide
Formula: C₈H₁₇N₃O
MW: 171.24 g/mol
Boiling Pt: 105…109 °C/0.2 mmHg
Storage Temperature: Ambient
MDL Number: MFCD01075232
CAS Number: 39890-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR