Benzylidenebis(tricyclohexylphosphine)dichlororuthenium 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Bis(tricyclohexylphosphine)benzylidine ruthenium(IV) dichloride

IN3049-1G IN3049-25G IN3049-5G
77401-544EA 64.18 USD
77401-544 77401-546 77401-548
Benzylidenebis(tricyclohexylphosphine)dichlororuthenium 95%
Benzylidenebis(tricyclohexylphosphine)dichlororuthenium
Formula: C₄₃H₇₂Cl₂P₂Ru
Melting Pt: 153 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01090946
CAS Number: 172222-30-9
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR