Calcium carbonate 99.99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
IN1381-100G
IN1381-25G
77401-386EA
467.64
USD
77401-386
77401-388
Calcium carbonate 99.99%
Calcium carbonate
Formula:
CaCO₃ Melting Pt: 800 °C (dec.) Density: 2.93 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010906 CAS Number: 471-34-1 EINECS: 207-439-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...