Calcium bromide, anhydrous 99,5%

Supplier: APOLLO SCIENTIFIC
Warning

IN1378-100G IN1378-250G
77401-382EA 103.92 USD
77401-382 77401-384
Calcium bromide, anhydrous 99,5%
Calcium bromide
Formula: Br₂Ca
Boiling Pt: 806…812 °C
Melting Pt: 730 °C (dec.)
Density: 3.353 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010902
CAS Number: 7789-41-5
EINECS: 232-164-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR