Bismuth(III) fluoride, anhydrous 99.99%

Supplier: APOLLO SCIENTIFIC
Danger

IN1243-100G IN1243-25G
77401-376EA 729.48 USD
77401-376 77401-378
Bismuth(III) fluoride, anhydrous 99.99%
Bismuth(III) fluoride
Formula: BiF₃
MW: 265.98
Boiling Pt: 230
Melting Pt: 727 °C
Density: 8.3 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010893
CAS Number: 7787-61-3
UN: 3260
ADR: 8 II
Merck Index: 12,01305

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR