2'-Nitroacetophenone 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR0043-10G OR0043-25G
77401-654EA 21.26 USD
77401-654 77401-656
2'-Nitroacetophenone 98%
2'-Nitroacetophenone
Formula: C₈H₇NO₃
Boiling Pt: 159 °C/16 mmHg
Melting Pt: 23…27 °C
Density: 1.24 g/cm³ (25 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007144
CAS Number: 577-59-3
EINECS: 209-414-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR