Oxalic acid dihydrate ACS
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Ethanedioic acid dihydrate
IN2794-5KG
IN2794-1KG
77401-514EA
597.06
USD
77401-514
77401-512
Oxalic acid dihydrate ACS
Oxalic acid dihydrate
Formula:
HO₂CCO₂H·2H₂O MW: 126.07 g/mol Melting Pt: 101…102 °C Density: 1.65 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149102 CAS Number: 6153-56-6 EINECS: 607-634-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...