Nickel(II) nitrate hexahydrate 99.995%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
IN2698-250G
IN2698-50G
77401-504EA
275.08
USD
77401-504
77401-506
Nickel(II) nitrate hexahydrate 99.995%
Nickel(II) nitrate hexahydrate
Formula:
Ni(NO₃)₂·6H₂O Boiling Pt: 137 °C Melting Pt: 56.7 °C Density: 2.05 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149805 CAS Number: 13478-00-7 EINECS: 236-068-5 UN: 2725 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...