Neodymium trichloride hexahydrate 99,9%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Neodymium (III) chloride hexahydrate
IN2632-100G
IN2632-50G
77401-496EA
234.32
USD
77401-496
77401-498
Neodymium trichloride hexahydrate 99,9%
Neodymium trichloride hexahydrate
Formula:
Cl₃Nd·6H₂O MW: 358.69 Melting Pt: 123 Density: 2.282 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149801 CAS Number: 13477-89-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...