Copper(II) sulfate pentahydrate 99%

Supplier: APOLLO SCIENTIFIC
Danger

IN1591-1KG IN1591-250G
77401-420EA 88.64 USD
77401-420 77401-422
Copper(II) sulfate pentahydrate 99%
Copper(II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
Melting Pt: 110 °C (dec.)
Density: 2.284 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR