L-(-)-tert-Leucine (H-Tle-OH) 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (S)-2-Amino-3,3-dimethylbutanoic acid, H-Tle-OH

OR0572-100G OR0572-25G OR0572-500G
77402-140EA 137.54 USD
77402-140 77402-142 77402-144
L-(-)-tert-Leucine (H-Tle-OH) 97%
L(-)-tert-Leucine (H-Tle-OH)
Formula: C₆H₁₃NO₂
MW: 131.2 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064218
CAS Number: 20859-02-3
EINECS: 424-750-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR