1-Isopropyl-1H-1,2,3-benzotriazole-5-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR0563-5G OR0563-1G OR0563-250MG
77402-134EA 268.96 USD
77402-134 77402-130 77402-132
1-Isopropyl-1H-1,2,3-benzotriazole-5-carboxylic acid
1-Isopropyl-1H-1,2,3-benzotriazole-5-carboxylic acid
Formula: C₁₀H₁₁N₃O₂
MW: 205.22 g/mol
Melting Pt: 222…231 °C
Storage Temperature: Ambient
MDL Number: MFCD01570664
CAS Number: 306935-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR