1,3-Dimethyl-1H-pyrazole-5-carbonitrile 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Cyano-1,3-dimethyl-1H-pyrazole

OR017148-250MG OR017148-1G
77401-814EA 22.42 USD
77401-814 77401-812
1,3-Dimethyl-1H-pyrazole-5-carbonitrile 95%
1,3-Dimethyl-1H-pyrazole-5-carbonitrile
Formula: C₆H₇N₃
MW: 121.14 g/mol
Melting Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00833260
CAS Number: 306936-77-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR