Silver carbonate 98%

Supplier: APOLLO SCIENTIFIC
Warning

IN3219-100G IN3219-25G IN3219-5G
77401-554EA 379 USD
77401-554 77401-556 77401-558
Silver carbonate 98%
Silver carbonate
Formula: CAg₂O₃
MW: 275.75 g/mol
Melting Pt: 218 °C (dec.)
Density: 6.08 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003403
CAS Number: 534-16-7
UN: 3077
ADR: 9+EHS III
Merck Index: 12,08650

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR