5-Methyl-4H-1,2,4-triazol-3-amine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR01571-1G OR01571-5G
77401-772EA 49.9 USD
77401-772 77401-774 77401-776
5-Methyl-4H-1,2,4-triazol-3-amine 98%
5-Methyl-4H-1,2,4-triazol-3-amine
Formula: C3H6N4
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00498236
CAS Number: 4923-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR