Ethyl-2-amino-5-methyl-4-phenylthiophene-3-carboxylate 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR0092-1G OR0092-250MG OR0092-5G
77401-680EA 52.98 USD
77401-680 77401-682 77401-684
Ethyl-2-amino-5-methyl-4-phenylthiophene-3-carboxylate 95%
Ethyl-2-amino-5-methyl-4-phenylthiophene-3-carboxylate
Formula: C₁₄H₁₅NO₂S
MW: 261.34
Melting Pt: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD01763475
CAS Number: 4815-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR