Titanium(IV) oxide rutile 99.99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Pigment White 6, Rutile, Titanium dioxide rutile
IN3706-25G
IN3706-100G
IN3706-250G
77401-594EA
75.4
USD
77401-594
77401-590
77401-592
Titanium(IV) oxide rutile 99.99%
Titanium(IV) oxide rutile
Formula:
O₂Ti MW: 79.88 g/mol Boiling Pt: 2500…3000 °C Melting Pt: 1855 Density: 4.26 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011269 CAS Number: 1317-80-2 EINECS: 215-282-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...