Lithium thiocyanate 99,9%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Lithium sulphocyanide, Lithium rhodanide

IN2380-100G IN2380-25G
77401-486EA 434.02 USD
77401-486 77401-488
Lithium thiocyanate 99,9%
Lithium thiocyanate
Formula: CLiNS
MW: 65.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011097
CAS Number: 556-65-0
EINECS: 209-135-1
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR