Copper(II) nitrate hemipentahydrate 99.999%

Supplier: APOLLO SCIENTIFIC

IN1570-100G IN1570-500G
77401-410EA 399.38 USD
77401-410 77401-412
Copper(II) nitrate hemipentahydrate 99.999%
Copper(II) nitrate hemipentahydrate
Formula: Cu(NO₃)₂·2.5H₂O
Melting Pt: 255 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02253001
CAS Number: 19004-19-4
UN: 3085
ADR: 5.1+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR