Copper(I) bromide dimethyl sulfide complex 99%

Supplier: APOLLO SCIENTIFIC
Warning

IN1541-10G IN1541-50G
77401-404EA 765.29 USD
77401-404 77401-406 77401-408
Copper(I) bromide dimethyl sulfide complex 99%
Copper (I) bromide dimethyl sulphide complex
Formula: C₂H₆BrCuS
Melting Pt: 132 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00043295
CAS Number: 54678-23-8
UN: 3335
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR