Cerium(IV) sulphate tetrahydrate 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Cerium disulphate tetrahydrate, Ceric sulphate tetrahydrate
IN1426-500G
IN1426-100G
77401-394EA
475.78
USD
77401-394
77401-392
Cerium(IV) sulphate tetrahydrate 99%
Cerium(IV) sulphate tetrahydrate
Formula:
Ce(SO₄)₂·4H₂O MW: 404.3 g/mol Density: 3.91 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149427 CAS Number: 10294-42-5 UN: 3085 ADR: 5.1+8 III Merck Index: 12,02036 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...