Pyridine-[D₅] (>99.50% D)

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Pyridine-[²H₅]

DE85BA-10ML
77401-320EA 262 USD
77401-320
Pyridine-[D₅] (>99.50% D)
Pyridine-[D5]

(HDO+D2O)<0.05%

Formula: C₅D₅N
MW: 84.06 g/mol
Boiling Pt: 114.4 °C
Density: 1.05 g/cm³ (25 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00044639
CAS Number: 7291-22-7
EINECS: 230-720-2
UN: 1282
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR