1-(Piperazin-1-yl)propan-1-one 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Propanoylpiperazine, 1-(1-Oxoprop-1-yl)piperazine

OR0854-1G OR0854-250MG
77402-270EA 36.2 USD
77402-270 77402-272
1-(Piperazin-1-yl)propan-1-one 98%
1-(Piperazin-1-yl)propan-1-one
Formula: C₇H₁₄N₂O
MW: 142.199 g/mol
Boiling Pt: 78 °C (0.2 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD04973346
CAS Number: 76816-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR