tert-Butyl [2-(methylamino)ethyl]carbamate hydrochloride 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR0802-250MG OR0802-5G OR0802-1G
77402-254EA 20.32 USD
77402-254 77402-256 77402-252
tert-Butyl [2-(methylamino)ethyl]carbamate hydrochloride 95%
tert-Butyl [2-(methylamino)ethyl]carbamate hydrochloride
Formula: C₈H₁₈N₂O₂·ClH
Melting Pt: 129…130 °C
Storage Temperature: Ambient
MDL Number: MFCD06796281
CAS Number: 202207-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR