Poly(ethylene glycol) dithiol, M.W. ~2000 97%

Supplier: Ambeed
Danger

A1483162-100MG A1483162-1G A1483162-250MG A1483162-5G
77441-318EA 21 USD
77441-318 77441-320 77441-322 77441-324
Poly(ethylene glycol) dithiol, M.W. ~2000 97%
Poly(ethylene glycol) dithiol

Precaución: for Research Use Only

Storage Temperature: Refrigerator
MDL Number: MFCD01867010
CAS Number: 68865-60-1
UN: 3335
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR