Precision Balances, Legal For Trade, Standard MA
Supplier: Mettler Toledo
30697485 30697476 30697479 30697482
77323-258EA 7184.55 USD
77323-258 77323-246 77323-250 77323-254
Precision Balances, Legal For Trade, Standard MA
Balances and Scales
METTLER TOLEDO Standard MA Legal for Trade Precision Balances have a large weighing pan, robust design and highly accurate weigh cell with internal adjustment.
  • Statistical data analysis
  • Front level indicator
  • Metal base, PBT top housing
  • Overload protection
  • Two interfaces: USB-A, RS232
  • Bluetooth option
The hybrid LCD touchscreen increases efficiency in the workflow with intuitive onscreen guidance and user management. Easily navigate 10 built-in applications and export weighing data. Built with robust construction to defend against external influences and provide high quality service for years to come. Easily export weighing data to a computer, printer, or USB storage device.
Certificaciones: OIML/NTEP Approved.