IntelliXcap™ septum caps streamlines high-throughput workflows, automating septum decapping, recapping, and capping. Ensures secure sealing, eliminating manual intervention and preserving sample integrity.
- Advanced automation for increased throughput
- Compact, modular design for ultimate flexibility
- No requirement for compressed air for flexible use within the lab
- Capping module available for Azenta labware, enabling capping of tubes from cap mat
- Single column harpoon decapping and recapping available as add-on
- Easy to use, easy to integrate; quick installation, easy-to-use touch panels bring instant control
- Wide operating temperature range of 15 to 35 °C
- Control is via a quick installation, easy-to-use touch panel
- Ideal for medium to high-throughput laboratories managing compound libraries, biobanks, or other biological samples.
- Suitable for use with tubes within a 96-format from Azenta as well as those from other manufacturers.
- Automation and speed reduces sample handling time, increasing sample throughput workflow.
This high-speed whole rack tube decapper and capper automatically removes, recaps, or disposes of septum caps from a full 96-format SBS rack of sample storage tubes. It can also be used to apply new sets of Azenta septum cap mats, without needing to use a separate instrument. Using the system for secure storage tube sealing removes the need for manual intervention and preserves sample integrity for use within high throughput environments such as in biobanking, compound libraries, and other storage applications.
Compatible with multiple sample storage tube types from a range of tube manufacturers, the system provides flexibility in either full automated robotic systems or as a standalone unit.