Stilla® naica® PCR MIX
Supplier: STILLA TECHNOLOGIES, INC.
R10056 R10106
77392-286EA 456.28 USD
77392-286 77392-292
Stilla® naica® PCR MIX
Nucleic Acid Reagents End-point PCR Enzymes and Kits
The naica® PCR MIX is a ready-to-use two-component solution optimized for EvaGreen® fluorescent intercalating dye-based Crystal Digital PCR®.
  • naica® PCR MIX 5X - Available in 1.50 ml volume size
  • naica® PCR MIX 10X - Available in 0.750 ml volume size
The naica® PCR MIX was specially-formulated to ensure optimal performance for EvaGreen® fluorescent intercalating dye-based Crystal Digital PCR® on the Nio™+ and naica® systems. The unique 10X concentration liberates a rection volume that can instead be utilized by additional sample input, which provides superior detection sensitivity for low concentration samples and rare targets.
Información de suministro: Dry-ice shipment.
Precaución: Store at −20 ±5 °C. For research use only. Not for use in diagnostic procedures.