Hemin chloride (from bovine) ≥90%, Sigma-Aldrich®

Synonyms: Chlorohemin, 1,3,5,8-Tetramethyl-2,4-divinylporphine-6,7-dipropionic acid ferrichloride, Chloroprotoporphyrin IX iron(III), Haemin, Hemin, Ferriprotoporphyrin IX chloride

M-Clarity™ quality level = MQ200

H9039-100G H9039-1G H9039-10G
220182-135EA 881.54 USD
220182-135 220182-137 220182-136
Hemin chloride (from bovine) ≥90%, Sigma-Aldrich®
Hemin chloride
Formula: C₃₄H₃₂ClFeN₄O₄
MW: 651.95 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00010726
CAS Number: 16009-13-5
EINECS: 240-140-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR