Hemin chloride (from porcine) ≥96.0% (by HPLC), BioXtra, Sigma-Aldrich®

Synonyms: Chlorohemin, 1,3,5,8-Tetramethyl-2,4-divinylporphine-6,7-dipropionic acid ferrichloride, Chloroprotoporphyrin IX iron(III), Haemin, Hemin, Ferriprotoporphyrin IX chloride

M-Clarity™ quality level = MQ200

51280-1G 51280-25G 51280-5G
220074-074EA 82.15 USD
220074-074 220074-075 220074-076
Hemin chloride (from porcine) ≥96.0% (by HPLC), BioXtra, Sigma-Aldrich®
Hemin chloride
Formula: C₃₄H₃₂ClFeN₄O₄
MW: 651.95 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00010726
CAS Number: 16009-13-5
EINECS: 240-140-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR