Sterilization Pouches, Heat Seal Tyvek®
Supplier: O&M HALYARD CANADA, INC CA
48528 48529 48530 48531 48532 48533 48534 48535 48536 48537
77322-670CS 310.34 USD
77322-670 77322-672 77322-674 77322-676 77322-678 77322-680 77322-682 77322-684 77322-686 77322-688 77322-690
Sterilization Pouches, Heat Seal Tyvek®
Bags Sterilization Pouches
HALYARD* Paper-Film single-use heat seal sterilization pouches are intended to be used to enclose another medical device, in a single or double pouch configuration, that is to be sterilized by a health care professional via steam and EO.
  • Single-use disposable
  • 68 GSM paper
Certificaciones: CE marked and comply with the international norms and standards ISO 11607 and EN 868.