ɑ-Caryophyllene, natural isolate

Supplier: Ricca Chemical

Synonyms: ɑ-Humulene, Humulene

RCCT9036-100C6 RCCT9036-20C6 RCCT9036-750C6
RCCT9036-100C6EA 170 USD
RCCT9036-100C6 RCCT9036-20C6 RCCT9036-750C6
ɑ-Caryophyllene, natural isolate
alpha-Caryophyllene
Formula: C₁₅H₂₄
MW: 204.36 g/mol
MDL Number: MFCD00042689
CAS Number: 6753-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR