(+/-)-ɑ-Bisabolol, natural isolate

Supplier: Ricca Chemical
Warning

Synonyms: rel-(R)-6-Methyl-2-((R)-4-methylcyclohex-3-en-1-yl)hept-5-en-2-ol, 6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol

RCCT9034-100C6 RCCT9034-20C6 RCCT9034-750C6
RCCT9034-100C6EA 357 USD
RCCT9034-100C6 RCCT9034-20C6 RCCT9034-750C6
(+/-)-ɑ-Bisabolol, natural isolate
(+/-)-alpha-Bisabolol
Formula: C₁₅H₂₆O
MW: 222.37 g/mol
MDL Number: MFCD00129082
CAS Number: 515-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR