(±)-α-Terpineol, natural isolate

Supplier: Ricca Chemical
Warning

Synonyms: α-Terpineol, Terpineol, 2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol

RCCT9031-100C6 RCCT9031-20C6 RCCT9031-750C6
RCCT9031-100C6EA 63 USD
RCCT9031-100C6 RCCT9031-20C6 RCCT9031-750C6
(±)-α-Terpineol, natural isolate
(±)-α-Terpineol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00001557
CAS Number: 98-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR