β-Caryophyllene, natural isolate

Supplier: Ricca Chemical
Danger

Synonyms: (1R,4E,9S)-4,11,11-trimethyl-8-methylene-bicyclo[7.2.0]undec-4-ene, (1R,9S,E)-4,11,11-Trimethyl-8-methylenebicyclo[7.2.0]undec-4-ene, (−)-β-Caryophyllene, (-)-trans-caryophyllene, NSC 11906

RCCT9030-100C6 RCCT9030-20C6 RCCT9030-750C6
RCCT9030-100C6EA 61 USD
RCCT9030-100C6 RCCT9030-20C6 RCCT9030-750C6
β-Caryophyllene, natural isolate
β-Caryophyllene
Formula: C₁₅H₂₄
MW: 204.36 g/mol
MDL Number: MFCD00075925
CAS Number: 87-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR